Mercury(I) nitrate dihydrate 97%

Danger

A16268.18 A16268.30
A16268.18EA 61.7 EUR
A16268.18 A16268.30
Mercury(I) nitrate dihydrate 97%
Mercury (I) nitrate dihydrate

Mercury(I) nitrate dihydrate is used in the preparation of other mercury(I) compounds. It acts as a reducing agent. It is involved in the preparation of mercury(II) nitrate by treating with dilute nitric acid as well as to get mercury on exposure to light.


Purity: 98% CAS No.: 14836-60-3 Molecular Formula: HgNO3.2H2O Formula Weight: 561.22 (525.19anhy) Odour: Odourless Melting Point: 70° UN No.: 1627 MDL No.: MFCD00150812

Formula: Hg₂N₂O₆·2H₂O
MW: 561.22 g/mol
Melting Pt: 70
Density: 4.79
Storage Temperature: Ambient
MDL Number: MFCD00150812
CAS Number: 7782-86-7
EINECS: 233-886-4
UN: 1627
Merck Index: 13,05923

Order Now

Specification Test Results

Appearance (Color) White to yellow to yellow-red
Form Crystalline or lumpy powder
Assay (unspecified) ≥96.0%
Comment May contain small amounts of dark particulate matter as part of manufacturing process.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR