(R)-(-)-2-Octanol, Supelco®

Warning


M-Clarity™ quality level = MQ100

147990-10G 147990-5G
147990-10GEA 410 EUR
147990-10G 147990-5G
(R)-(-)-2-Octanol, Supelco®
(R)-(-)-2-Octanol
Formula: C₈H₁₈O
Boiling Pt: 175 °C (1013 hPa)
Density: 0,8201 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064284
CAS Number: 5978-70-1
EINECS: 227-777-0
UN: 1987
ADR: 3,III
Merck Index: 12,06850
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=18fd46b3d9d8ab0ee6af423314fb7b52b40bdfc5&vid=18fd46b3d9d8ab0ee6af423314fb7b52b40bdfc5&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR