(R)-(+)-α-Methylbenzylamine, Supelco®
Supplier: SUPELCO
|
Danger
|
M-Clarity™ quality level = MQ100
77879-25ML
77879-5ML
77879-25MLEA
367
EUR
77879-25ML
77879-5ML
(R)-(+)-α-Methylbenzylamine, Supelco®
(R)-(+)-α-Methylbenzylamine
Formula:
C₈H₁₁N Boiling Pt: 180 °C (1013 hPa) Melting Pt: 32,5 °C Density: 0,95 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064405 CAS Number: 3886-69-9 EINECS: 223-423-4 Merck Index: 12,06106 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...