2-Bromoethanol 97%
Supplier: ACROS ORGANICS
|
Danger
|
106920050
106921000
106922500
ACRO106920050EA
68.2
EUR
ACRO106920050
ACRO106921000
ACRO106922500
2-Bromoethanol 97%
2-Bromoethanol
Formula:
BrH₂CCH₂OH Boiling Pt: 149…150 °C (750 mmHg) Melting Pt: –80 °C Density: 1,76 g/cm³ Flash Pt: >110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00002827 CAS Number: 540-51-2 EINECS: 208-748-1 UN: 2927 ADR: 6.1,II Merck Index: 13,03826 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid (May darken on storage) |
Infrared spectrum | Conforms |
Refractive index | 1.4890 to 1.4940 (20°C, 589 nm) |
GC | ≥98.5 % |
Acid value | ≤0.1 mg KOH/g |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...