Methyl difluoroacetate ≥98%
|
Danger
|
B22197.06
B22197.14
B22197.06EA
52.1
EUR
B22197.06
B22197.14
Methyl difluoroacetate ≥98%
Methyl difluoroacetate
Purity: 99%
CAS No.: 433-53-4
Molecular Formula: F2CHCO2CH3
Formula Weight: 110.06
Flash Point: 19°(66°F)
Refractive Index: 1.3350
Boiling Point: 86-87°
Beilstein Reference: 1747534
UN No.: 2924
MDL No.: MFCD00039276
Formula:
C₃H₄F₂O₂ Boiling Pt: 86…87 °C Density: 1,272 Flash Pt: 19 °C (66 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039276 CAS Number: 433-53-4 EINECS: 207-089-7 UN: 2924 ADR: 3,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...