1,6-Diaminohexane for synthesis, Sigma-Aldrich®

Danger


M-Clarity™ quality level = MQ200

8.04323.1000 8.04323.9025
8.04323.1000EA 99.3 EUR
8.04323.1000 8.04323.9025
1,6-Diaminohexane for synthesis, Sigma-Aldrich®
1,6-Diaminohexane
Formula: C₆H₁₆N₂
MW: 116.21 g/mol
Boiling Pt: 205
Melting Pt: 39…42 °C
Density: 0.66 g/cm³ (20 °C)
Flash Pt: 80
Storage Temperature: Ambient
MDL Number: MFCD00008243
CAS Number: 124-09-4
EINECS: 204-679-6
UN: 2280
ADR: 8 III
Merck Index: 13,04713

Order Now

Specification Test Results

Solubility 490 g/l (20 °C)
Melting Point 39 - 42 °C
Molar Mass 116.21 g/mol
Boiling Point 199 - 204 °C (1013 hPa)
Vapor Pressure 0.25 hPa (20 °C)
Flash Point 85 °C
Explosion Limit 0.9 - 7.6 %(V)
Density 0.83 g/cm3 (60 °C)
pH 12.4 (100 g/l, H2O, 25 °C)
Ignition Point 305 °C
Hazard Class 8
Hazard Class Description Corrosive Material


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR