1-Cyclohexylpiperidine ≥97%
Supplier: ALFA AESAR
|
Warning
|
L07897.09
L07897.18
L07897.09EA
0
EUR
L07897.09
L07897.18
1-Cyclohexylpiperidine ≥97%
1-Cyclohexylpiperidine
Purity: 97%
CAS No.: 3319-01-5
Molecular Formula: C11H21N
Formula Weight: 167.30
Refractive Index: 1.4880
Boiling Point: 231-234°
MDL No.: MFCD00014642
Formula:
C₁₁H₂₁N Boiling Pt: 231…234 °C Density: 0,914 Storage Temperature: Ambient |
MDL Number:
MFCD00014642 CAS Number: 3319-01-5 EINECS: 222-016-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...