4-Bromo-3,5-difluoroanisole ≥98%

Warning

A17427.06 A17427.14
A17427.06EA 34.5 EUR
A17427.06 A17427.14
4-Bromo-3,5-difluoroanisole ≥98%
4-Bromo-3,5-difluoroanisole

Purity: 98% CAS No.: 202865-61-0 Molecular Formula: C7H5BrF2O Formula Weight: 223.02 Refractive Index: 1.5228 Melting Point: 26-30° MDL No.: MFCD00143268

Formula: C₇H₅BrF₂O
Melting Pt: 26…30 °C
Storage Temperature: Ambient
MDL Number: MFCD00143268
CAS Number: 202865-61-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR