3,4-Dichlorobenzamide ≥97%
Supplier: ALFA AESAR
A13878.03
A13878.06
A13878.03EA
0
EUR
A13878.03
A13878.06
3,4-Dichlorobenzamide ≥97%
3,4-Dichlorobenzamide
Purity: 97%
CAS No.: 2670-38-4
Molecular Formula: C7H5Cl2NO
Formula Weight: 190.03
Melting Point: 140-142°
Beilstein Reference: 3089065
Formula:
C₇H₅Cl₂NO Melting Pt: 140…142 °C Storage Temperature: Ambient |
MDL Number:
MFCD00053009 CAS Number: 2670-38-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...