Diphenyl ether, Supelco®
Supplier: SUPELCO
|
|
M-Clarity™ quality level = MQ100
67334-25ML
67334-5ML
67334-25MLEA
51.1
EUR
67334-25ML
67334-5ML
Diphenyl ether, Supelco®
Diphenyl ether
Formula:
C₁₂H₁₀O Boiling Pt: 498 °C (1013 hPa) Melting Pt: 24…27 °C Density: 1,07 g/cm³ (20 °C) Flash Pt: 125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003034 CAS Number: 101-84-8 EINECS: 202-981-2 UN: 3077 ADR: 9,III Merck Index: 12,07442 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...