(3-Chlorophenyl)acetonitrile ≥99%
|
Danger
|
A15673.06
A15673.14
A15673.22
A15673.06EA
22.2
EUR
A15673.06
A15673.14
A15673.22
(3-Chlorophenyl)acetonitrile ≥99%
(3-Chlorophenyl)acetonitrile
Purity: 99%
CAS No.: 1529-41-5
Molecular Formula: C8H6ClN
Formula Weight: 151.60
Flash Point:
Formula:
C₈H₆ClN Boiling Pt: 134…136 °C (10 mmHg) Melting Pt: 11…13 °C Density: 1,189 Flash Pt: >110 °C (230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001908 CAS Number: 1529-41-5 EINECS: 216-213-9 UN: 3276 ADR: 6.1,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...