Cyclohexylméthanol ≥99%

Supplier: ALFA AESAR
Warning

L06538.14 L06538.22
L06538.14EA 55.7 EUR
L06538.14 L06538.22
Cyclohexylméthanol ≥99%
Cyclohexylméthanol

Purity: 99% CAS No.: 100-49-2 Molecular Formula: C7H14O Formula Weight: 114.19 Flash Point: 71°(159°F) Refractive Index: 1.4640 Boiling Point: 180-181° Beilstein Reference: 773712 Merck Reference: 14,2731 MDL No.: MFCD00001510

Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 180…181 °C
Melting Pt: –43 °C
Density: 0.926
Flash Pt: 71 °C (159 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001510
CAS Number: 100-49-2
EINECS: 202-857-8
Merck Index: 13,02760

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR