Cesium bromide ≥99.999% (metals basis)
|
Warning
|
87639.09
87639.18
87639.09EA
153
EUR
87639.09
87639.18
Cesium bromide ≥99.999% (metals basis)
Cesium bromide
Purity: 99.999%
CAS No.: 7787-69-1
Molecular Formula: BrCs
Formula Weight: 212.81
Physical Form: -4 Mesh
Density: 4.44
Refractive Index: 1.6984
Sensitivity: Hygroscopic
Appearance: White
Odour: Odourless
Melting Point: 636°
Boiling Point: 1300°
Merck Reference: 14,2009
MDL No.: MFCD00010954
Formula:
BrCs MW: 212.81 g/mol Boiling Pt: 1300 °C Melting Pt: 636 Density: 4.44 Storage Temperature: Ambient |
MDL Number:
MFCD00010954 CAS Number: 7787-69-1 EINECS: 232-130-0 Merck Index: 14,2009 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...