Lithium niobate 99%

A17805.06 A17805.18 A17805.30
A17805.06EA 46.4 EUR
A17805.06 A17805.18 A17805.30
Lithium niobate 99%
Lithium niobate

Purity: 99.99% CAS No.: 12031-63-9 Molecular Formula: LiNbO3 Formula Weight: 147.85 Odour: Odourless Melting Point: ca 1275° MDL No.: MFCD00011080

Formula: LiNbO₃
MW: 147.85 g/mol
Melting Pt: ∼1275 °C
Density: 4.3
Storage Temperature: Ambient
MDL Number: MFCD00011080
CAS Number: 12031-63-9
EINECS: 234-755-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR