Lead(II) bromide, ultra dry ≥99.999% (metals basis)
Supplier: ALFA AESAR
|
Danger
|
ampouled under argon
35703.03
35703.06
35703.14
35703.03EA
122
EUR
35703.03
35703.06
35703.14
Lead(II) bromide, ultra dry ≥99.999% (metals basis)
Lead (II) bromide
Purity: 99.999%
CAS No.: 10031-22-8
Molecular Formula: Br2Pb
Formula Weight: 367.01
Physical Form: -10 Mesh Beads, Ampouled under argon
Density: 6.66
Appearance: White
Odour: Odourless
Melting Point: 373°
Boiling Point: 916°
Merck Reference: 14,5403
UN No.: 2291
MDL No.: MFCD00011156
Formula:
Br₂Pb Boiling Pt: 916 °C Melting Pt: 373 °C Density: 6,66 Storage Temperature: Ambient |
MDL Number:
MFCD00011156 CAS Number: 10031-22-8 EINECS: 233-084-4 UN: 2291 ADR: 6.1,III Merck Index: 14,5403 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...