Antimony(III) fluoride for synthesis, Sigma-Aldrich®

Supplier: MERCK MILLIPORE
Danger


M-Clarity™ quality level = MQ200

8.07839.0005 8.07839.0250
8.07839.0005EA 25.6 EUR
8.07839.0005 8.07839.0250
Antimony(III) fluoride for synthesis, Sigma-Aldrich®
Antimony (III) fluoride
Formula: F₃Sb
Boiling Pt: 376 °C (1013 hPa)
Melting Pt: 292 °C
Density: 4,38 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011218
CAS Number: 7783-56-4
EINECS: 232-009-2
UN: 1549
ADR: 6.1,III
Merck Index: 13,00715

Order Now

Specification Test Results

Solubility 4430 g/l (20 °C) Hydrolysis
Melting Point 292 °C
Molar Mass 178.74 g/mol
Bulk Density 1500 kg/m3
Boiling Point 376 °C (1013 hPa)
Density 4.38 g/cm3 (20 °C)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR