Tributyl phosphate, Supelco®

Supplier: SUPELCO
Warning


M-Clarity™ quality level = MQ100

00675-100ML 00675-500ML
00675-100MLEA 42.9 EUR
00675-100ML 00675-500ML
Tributyl phosphate, Supelco®
Tributyl phosphate
Formula: (CH₃(CH₂)₃O)₃PO
Boiling Pt: 289 °C (1013 hPa)
Melting Pt: –79 °C
Density: 0,982 g/cm³ (20 °C)
Flash Pt: 151 °C
Storage Temperature: Ambient
MDL Number: MFCD00009436
CAS Number: 126-73-8
EINECS: 204-800-2
Merck Index: 13,09692
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=83c36cd3c4efe2452f97c8983300d14886417d08&vid=83c36cd3c4efe2452f97c8983300d14886417d08&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR