Dimethyl sulphoxide

Supplier: ROTH CARL

A994.1 A994.2
ROTHA994.1EA 35.5 EUR
ROTHA994.1 ROTHA994.2
Dimethyl sulphoxide
Dimethyl sulphoxide
Formula: (CH₃)₂SO
Boiling Pt: 189 °C (1013 hPa)
Melting Pt: 18,5 °C
Density: 1,101 g/cm³ (20 °C)
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00002089
CAS Number: 67-68-5
EINECS: 200-664-3
Merck Index: 13,03285
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3ec1573e80c11ddf9254283ef76abf5fb6539c7e&vid=d0727d3220885f6adab5f099d9fd3b4de38d5710&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR