2-Ethylimidazole 99%
Supplier: ACROS ORGANICS
|
Danger
|
219281000
219285000
219280025
ACRO219281000EA
47.9
EUR
ACRO219281000
ACRO219285000
ACRO219280025
2-Ethylimidazole 99%
2-Ethylimidazole
Formula:
C₅H₈N₂ Boiling Pt: 268 °C (760 mmHg) Melting Pt: 83…87 °C Density: 1,03 g/cm³ (25 °C) Flash Pt: 159 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005192 CAS Number: 1072-62-4 EINECS: 214-011-5 |
Specification Test Results
Appearance | White to light yellow Flakes |
Infrared spectrum | Conforms |
Melting point | 83 to 87 °C |
Water | ≤0.5 % |
GC | ≤1.0 % Imidazole |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...