3-(tert-Butyldimethylsiloxy)glutaric anhydride 95%
|
Warning
|
H61227.14
H61227.06
H61227.14EA
288
EUR
H61227.14
H61227.06
3-(tert-Butyldimethylsiloxy)glutaric anhydride 95%
3-(tert-Butyldimethylsiloxy)glutaric anhydride
Formula:
C₁₁H₂₀O₄Si Melting Pt: 79…81 °C Density: 1,030 Flash Pt: >113 °C (235 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00075235 CAS Number: 91424-40-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...