L(-)-Tryptophan 98.5-101.5% (dry basis)
J22765.A1
J22765.22
J22765.A1EA
865
EUR
J22765.A1
J22765.22
L(-)-Tryptophan 98.5-101.5% (dry basis)
L(-)-Tryptophan
Formula:
C₁₁H₁₂N₂O₂ MW: 204.23 g/mol Melting Pt: 280…285 °C Density: 1.34 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00064340 CAS Number: 73-22-3 EINECS: 200-795-6 Merck Index: 14,09797 |
Specification Test Results
Assay (Dry Basis) | 98.5 to 101.5% |
Chloride Content | ≤0.05% |
Identification (FTIR) | Conforms |
Residue on Ignition | ≤0.1% |
Impurity Content, Organic Impurities | Meets the Requirements |
Impurity Content Sulphate (SO₄) | ≤0.03% |
Loss on Drying | ≤0.3% |
Optical Rotation | –29.4 to –32.8° (c=1 in H₂O) |
pH (1%, H₂O) | 5.5 to 7.0 |
Heavy Metals (Pb) | ≤15 ppm |
Iron (Fe) | ≤0.003% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...