Dimethyl adipate for synthesis


M-Clarity™ quality level = MQ200

8.44004.0100 8.44004.1000
8.44004.0100EA 25.7 EUR
8.44004.0100 8.44004.1000
Dimethyl adipate for synthesis
Dimethyl adipate
Formula: C₈H₁₄O₄
MW: 174.2 g/mol
Boiling Pt: 215…225 °C (1013 hPa)
Melting Pt: 10.3
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 107 °C
MDL Number: MFCD00008469
CAS Number: 627-93-0
Merck Index: 13,00163

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR