High temperature controlled atmosphere ovens, HTMA
Supplier: CARBOLITE
HTMA6220-230SN HTMA6220-400TN HTMA6028-230SN
CARBHTMA6220-230SNEA 23010 EUR
CARBHTMA6220-230SN CARBHTMA6220-400TN CARBHTMA6028-230SN
High temperature controlled atmosphere ovens, HTMA
Ovens High Temperature Ovens
The HTMA series is a range of modified atmosphere high temperature ovens for use with inert atmospheres up to 700 °C.
  • Digital 301 controller with single ramp to set-point and process timer
  • Rear mounted fan and side air guides give horizontal airflow
  • Fully seam welded to contain modified atmosphere
  • Manual gas control via needle valves and flowmeters
  • Single side hinged door, with metal heat seal and rubber gas tight seal, closed using non slam handle
Separate flow controls for purge and process gases mean that once the chamber has been purged of atmospheric air process gas can be used with lower flow rates. Switching between purge and process gases can either be done manually or by adding the option of an automatic programmable control system. Oxygen levels down to 50 ppm are achievable.
Interior made of corrosion resistant stainless steel with perforated non-tip shelves and runners. Features stainless steel pipe-work, nickel brass flow-meter and solenoid valves. Exterior made of hard wearing, zinc coated and stoved epoxy polyester coated. Gas inlet connection: bulk head compression fitting to suit 10 mm outside diameter tube (maximum inlet pressure = 2 bar).
Ordering information: Options available include a range of sophisticated digital controllers, multi-segment programmers and data loggers, stainless steel flow-meter and solenoid valves, instead of nickel brass, fixed or castor mounted floor stands, flow failure alarm, oxygen sensor fitted to gas outlet to monitor oxygen level. Only available for use with nanodac instruments.