Methyl trifluoromethanesulphonate 98% (by GC), LiChropur™ for GC derivatisation, Supelco®

18503-1G 18503-5G
18503-1GEA 38.8 EUR
18503-1G 18503-5G
Methyl trifluoromethanesulphonate 98% (by GC), LiChropur™ for GC derivatisation, Supelco®
Methyl trifluoromethanesulphonate
Formula: C₂H₃F₃O₃S
MW: 164.11 g/mol
Boiling Pt: 95
Density: 1.45
MDL Number: MFCD00000409
CAS Number: 333-27-7
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c38f531bc4d81c686782ef296defe546231b52a4&vid=c38f531bc4d81c686782ef296defe546231b52a4&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR