Organic reference standard, (R)-2-Methyl-3-ureidopropionic acid, Supelco®
06997-10MG
06997-50MG
06997-10MGEA
130
EUR
06997-10MG
06997-50MG
Organic reference standard, (R)-2-Methyl-3-ureidopropionic acid, Supelco®
Standards
Organic Standards
This metabolite is increased in the urine of patients with beta-ureidopropionase deficiency and can be used to predict patient′s individual phenotypes of enzyme deficiencies in pyrimidine metabolism when associated with a risk for severe toxicity against the antineoplastic agent 5-fluorouracil.