Organic reference standard, N-Cinnamoylglycine, analytical standard, Supelco®
Supplier: SUPELCO
98956-10MG 98956-50MG
98956-10MGEA 85.7 EUR
98956-10MG 98956-50MG
Organic reference standard, N-Cinnamoylglycine, analytical standard, Supelco®
Standards Organic Standards
Cinnamoylglycine is known as a urinary metabolite in man, although whether it is formed de novo from plant cinnamate or is a plant product excreted unchanged has not been conclusively demonstrated. When cinnamoylglycine occurs naturally it is probably a food constituent excreted unchanged. It is not found when small quantities (0,5 to 6 g) of cinnamic acid are fed to man, but by analogy with animal experiments may be produced when much larger quantities are given.