Organic reference standard, N-Decanoylglycine, analytical standard, Supelco®
93204-10MG 93204-250MG 93204-50MG
93204-10MGEA 129 EUR
93204-10MG 93204-250MG 93204-50MG
Organic reference standard, N-Decanoylglycine, analytical standard, Supelco®
Standards Organic Standards
Decanoylglycine is an acylglycine with C-10 fatty acid group as the acyl moiety. Acylglycines possess a common amidoacetic acid moiety and are normally minor metabolites of fatty acids. Elevated levels of certain acylglycines appear in the urine and blood of patients with various fatty acid oxidation disorders.
They are normally produced through the action of glycine N-acyltransferase which is an enzyme.