Sigma Pseudo™ corpse scent, drowned victim scent, Supelco®
Supplier: SUPELCO
P7184-15CAP P7184-50CAP
P7184-15CAPEA 57.8 EUR
P7184-15CAP P7184-50CAP
Sigma Pseudo™ corpse scent, drowned victim scent, Supelco®
Standards Organic Standards
A valuable training aid for water search. It provides a reliable scent source for 30 to 45 minutes, in still or running water, at depths of 1 to 12 feet. Canine training aids for the detection of corpses.
Packaging: 15, 50 caps in poly bottle.