Mercury ≥99.5%, AnalaR NORMAPUR® for polarography, tridistilled
|
Danger
|
85967.180EA
111
EUR
85967.180
85967.260
Mercury ≥99.5%, AnalaR NORMAPUR® for polarography, tridistilled
Mercury
Formula:
Hg MW: 200.59 g/mol Boiling Pt: 356.5 Melting Pt: –39 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011035 CAS Number: 7439-97-6 UN: 2809 ADR: 8 III Merck Index: 13,05925 |
Specification Test Results
Assay | Min. 99,5 % |
Matter insoluble in HNO3 | Max. 20 ppm |
Ag (Silver) | Max. 5 ppm |
Bi (Bismuth) | Max. 0,1 ppm |
Cd (Cadmium) | Max. 0,1 ppm |
Cu (Copper) | Max. 0,1 ppm |
Fe (Iron) | Max. 0,5 ppm |
Ni (Nickel) | Max. 0,1 ppm |
Pb (Lead) | Max. 0,1 ppm |
Tl (Thallium) | Max. 0,5 ppm |
Zn (Zinc) | Max. 0,5 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...