Boc-L-Cys-OH ≥98.5%, LiChropur™ for chiral derivatisation, Supelco®


M-Clarity™ quality level = MQ100

15411-1G 15411-250MG
15411-1GEA 251 EUR
15411-1G 15411-250MG
Boc-L-Cys-OH ≥98.5%, LiChropur™ for chiral derivatisation, Supelco®
Boc-L-Cys-OH

Chiral derivatizing agent used together with OPA for assaying the enantiomeric purity of amino acids


Boc-L-cysteine (Boc-Cys-OH) is a high performance liquid chromatography (HPLC) grade chromatographic solvent.


Optical activity: [α]20/D +8.0±1.5°, c = 1% in ethanol
Optical purity: Enantiomeric ratio: ≥99.5:0.5

Formula: C₈H₁₅NO₄S
MDL Number: MFCD00065565
CAS Number: 20887-95-0
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9dc81cfbc859ad6e4655bd56d77443d5c52f987f&vid=9dc81cfbc859ad6e4655bd56d77443d5c52f987f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR