4,5-Dimethoxy-o-phenylenediamine dihydrochloride ≥98.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
36271-100MG
36271-500MG
36271-100MGEA
81.1
EUR
36271-100MG
36271-500MG
4,5-Dimethoxy-o-phenylenediamine dihydrochloride ≥98.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
4,5-Dimethoxy-o-phenylenediamine dihydrochloride
Formula:
C₈H₁₂N₂O₂·2HCl MW: 241.12 g/mol |
MDL Number:
MFCD00190674 CAS Number: 131076-14-7 |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...