Poly(vinyl chloride), carboxylated 10% carboxyl basis, Selectophore™ ionophore, Supelco®

Warning

50133-10G 50133-1G
50133-10GEA 2760 EUR
50133-10G 50133-1G
Poly(vinyl chloride), carboxylated 10% carboxyl basis, Selectophore™ ionophore, Supelco®
Poly(vinyl chloride), carboxylated
Formula: (C₅H₇ClO₂)n
MDL Number: MFCD00084460
CAS Number: 25702-80-1
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b97858b1347dfa66549086de138f3ff781fb0110&vid=b97858b1347dfa66549086de138f3ff781fb0110&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR