2-(Aminomethyl)pyridine ≥98.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®

Danger

65562-10ML 65562-1ML
65562-10MLEA 51.8 EUR
65562-10ML 65562-1ML
2-(Aminomethyl)pyridine ≥98.0% (by GC), LiChropur™ derivatisation reagent, for HPLC, Supelco®
2-(Aminomethyl)pyridine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 197
Melting Pt: 81
MDL Number: MFCD00006360
CAS Number: 3731-51-9
UN: 2735
ADR: 8 II
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f859d9cff95088da7d55c498248ecdff5920225&vid=7f859d9cff95088da7d55c498248ecdff5920225&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR