G418 disulphate, Ultra Pure Grade
|
Warning
|
E859-1GEA
75.1
EUR
E859-1G
E859-5G
G418 disulphate, Ultra Pure Grade
G418 disulphate
Selection agent for both prokaryotic and eukaryotic transfected cells. An aminoglycoside similar to gentamycin, G418 is toxic to bacterial, yeast, higher plant and mammalian cells, protozoans, and helminths.
Formula:
C₂₀H₄₄N₄O₁₈S₂ Storage Temperature: Ambient |
MDL Number:
MFCD00058314 CAS Number: 108321-42-2 |
Specification Test Results
Activity | 500 U/mg |
Appearance | PASS |
Expiration Date | REPORT |
Identification (IR) | PASS |
Moisture | 10.0 % |
Optical Rotation (1%, Water)@25 °C | REPORT |
Solubility (80 mg/ml, PBS) | NONE |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...