Glycerine 86%

Supplier: ROTH CARL

4043.1 4043.2 4043.3
ROTH4043.1EA 0 EUR
ROTH4043.1 ROTH4043.2 ROTH4043.3
Glycerine 86%
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
MW: 92.09 g/mol
Boiling Pt: 290
Melting Pt: 18.6
Flash Pt: 177
Storage Temperature: Ambient
MDL Number: MFCD00004722
CAS Number: 56-81-5
EINECS: 200-289-5
Merck Index: 13,04497
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=481dd90ecbdec73cd4020432d3356161ebfe7681&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR