Calcium acetate monohydrate ≥99.0% ACS, Fluka™
Supplier: HONEYWELL
402850-100G
402850-500G
402850-100GEA
82.86
EUR
402850-100G
402850-500G
Calcium acetate monohydrate ≥99.0% ACS, Fluka™
Calcium acetate monohydrate
Formula:
(H₃CCOO)₂Ca·1H₂O MW: 176.18 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00150019 CAS Number: 5743-26-0 EINECS: 200-540-9 Merck Index: 13,01645 |
Specification Test Results
Assay | 99.0 - 106 % |
Titrable Acid | Max. 0.035 meq/g |
Alkalinity | complying |
Water insoluble matter | Max. 0.005 % |
Barium (Ba) | Max. 0.01 % |
Iron (Fe) | Max. 0.001 % |
Potassium (K) | Max. 0.01 % |
Magnesium (Mg) | Max. 0.05 % |
Sodium (Na) | Max. 0.02 % |
Strontium (Sr) | Max. 0.05 % |
Heavy metals (as Pb) | Max. 0.005 % |
Chloride (Cl) | Max. 0.001 % |
Sulfate (SO₄) | Max. 0.01 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...