Digallium trioxide ≥99.995% (metals basis)
36318.06
36318.14
36318.06EA
73.7
EUR
36318.06
36318.14
Digallium trioxide ≥99.995% (metals basis)
Digallium trioxide
Purity: 99.995%
CAS No.: 12024-21-4
Molecular Formula: Ga2O3
Formula Weight: 187.44
Physical Form: 3-12mm Sintered Lump
Density: 6.44
Refractive Index: 1.92
Odour: Odourless
Melting Point: 1900°
Merck Reference: 14,4346
MDL No.: MFCD00011020
For vacuum deposition
Formula:
Ga₂O₃ MW: 187.44 g/mol Melting Pt: 1900 °C Density: 6.44 Storage Temperature: Ambient |
MDL Number:
MFCD00011020 CAS Number: 12024-21-4 EINECS: 234-691-7 Merck Index: 14,4346 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...