Bismuth(III) oxide ≥99.5%, NanoArc®
|
Warning
|
Purity: 99.5+%
CAS No.: 1304-76-3
Molecular Formula: Bi2O3
Formula Weight: 465.96
Physical Form: 38nm APS Powder, S.A. 18m2/g
Density: 8.9
Appearance: Bright yellow
Odour: Odourless
Melting Point: 817°
Boiling Point: ca 1890°
Merck Reference: 14,1273
MDL No.: MFCD00003462
X-ray opaque particle. NanoArc™ bismuth oxide can be compounded into polymers or coating systems to increase opacity to x-rays,while the small particle size ensures minimal disruption of physical properties.
Formula:
BiO₃ MW: 465.96 g/mol Boiling Pt: ∼1890 °C Melting Pt: 817 °C Density: 8.9 Storage Temperature: Ambient |
MDL Number:
MFCD00003462 CAS Number: 1304-76-3 EINECS: 215-134-7 Merck Index: 14,1273 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...