Silodosin

Supplier: ACROS ORGANICS
Warning

464521000 464525000
ACRO464521000EA 74.3 EUR
ACRO464521000 ACRO464525000
Silodosin
Silodosin

  • Bioactive

Formula: C₂₅H₃₂F₃N₃O₄
MW: 495.54 g/mol
Melting Pt: >104 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00930170
CAS Number: 160970-54-7

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.3 %
HPLC ≥97.5 %
Residue after ignition ≤0.1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR