3,3,3-Trichloroprop-1-ene 95%

Supplier: Apollo Scientific
Warning

OR16695-25G OR16695-5G
APOSOR16695-25GEA 395 EUR
APOSOR16695-25G APOSOR16695-5G
3,3,3-Trichloroprop-1-ene 95%
3,3,3-Trichloroprop-1-ene
Formula: C₃H₃Cl₃
Boiling Pt: 114,5 °C
Melting Pt: –30 °C
Density: 1,33 g/cm³ (25 °C)
Storage Temperature: Freezer
MDL Number: MFCD01749212
CAS Number: 2233-00-3
UN: 1993
ADR: 3,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR