Methyl 3-(hydroxymethyl)cubane-1-carboxylate

OR312522-1G OR312522-250MG
APOSOR312522-1GEA 3730 EUR
APOSOR312522-1G APOSOR312522-250MG
Methyl 3-(hydroxymethyl)cubane-1-carboxylate
Methyl 3-(hydroxymethyl)cubane-1-carboxylate
Formula: C₁₁H₁₂O₃
Storage Temperature: Ambient
MDL Number: MFCD29075620
CAS Number: 1980053-96-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR