(4aR,7aS)-4-Benzyloctahydropyrrolo[3,4-b][1,4]oxazine ≥98%

Supplier: Apollo Scientific

OR361746-100MG OR361746-1G OR361746-250MG
APOSOR361746-100MGEA 287 EUR
APOSOR361746-100MG APOSOR361746-1G APOSOR361746-250MG
(4aR,7aS)-4-Benzyloctahydropyrrolo[3,4-b][1,4]oxazine ≥98%
(4aR,7aS)-4-Benzyloctahydropyrrolo[3,4-b][1,4]oxazine
Formula: C₁₃H₁₈N₂O
Storage Temperature: Ambient
MDL Number: MFCD28396738
CAS Number: 151213-50-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR