(S)-Cyclobutyl(phenyl)methanamine hydrochloride ≥98%

Supplier: Apollo Scientific

OR370007-100MG OR370007-1G OR370007-250MG
APOSOR370007-100MGEA 94.4 EUR
APOSOR370007-100MG APOSOR370007-1G APOSOR370007-250MG
(S)-Cyclobutyl(phenyl)methanamine hydrochloride ≥98%
(S)-Cyclobutyl(phenyl)methanamine hydrochloride
Formula: C₁₁H₁₅N·ClH
Storage Temperature: Ambient
MDL Number: MFCD20691543
CAS Number: 1202478-42-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR