(R)-N-Boc-2-aminobutylamine 97%
Supplier: Apollo Scientific
|
Warning
|
OR452197-1G
OR452197-250MG
OR452197-500MG
APOSOR452197-1GEA
654
EUR
APOSOR452197-1G
APOSOR452197-250MG
APOSOR452197-500MG
(R)-N-Boc-2-aminobutylamine 97%
(R)-N-Boc-2-aminobutylamine
Formula:
C₉H₂₀N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD22397599 CAS Number: 1194975-22-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...