4-Ethoxypicoline 98%

Supplier: Apollo Scientific

OR909512-10G OR909512-1G OR909512-5G
APOSOR909512-10GEA 754 EUR
APOSOR909512-10G APOSOR909512-1G APOSOR909512-5G
4-Ethoxypicoline 98%
4-Ethoxypicoline
Formula: C₈H₁₁NO
MW: 137.179 g/mol
Storage Temperature: Ambient
MDL Number: MFCD20489568
CAS Number: 18615-87-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR