2-Piperidinothiazole 98%
Supplier: Apollo Scientific
OR910251-1G
OR910251-5G
APOSOR910251-1GEA
219
EUR
APOSOR910251-1G
APOSOR910251-5G
2-Piperidinothiazole 98%
2-Piperidinothiazole
Formula:
C₈H₁₂N₂S MW: 168.259 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00234458 CAS Number: 4175-70-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...