2-(2-Methylphenyl)benzonitrile 96%

Supplier: Apollo Scientific

OR910704-1G OR910704-5G
APOSOR910704-1GEA 219 EUR
APOSOR910704-1G APOSOR910704-5G
2-(2-Methylphenyl)benzonitrile 96%
2-(2-Methylphenyl)benzonitrile
Formula: C₁₄H₁₁N
MW: 193.244 g/mol
Storage Temperature: Ambient
MDL Number: MFCD05980950
CAS Number: 157366-46-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR