N-Ethylthiophene-3-carboxamide 98%

Supplier: Apollo Scientific

OR911202-1G OR911202-5G
APOSOR911202-1GEA 219 EUR
APOSOR911202-1G APOSOR911202-5G
N-Ethylthiophene-3-carboxamide 98%
N-Ethylthiophene-3-carboxamide
Formula: C₇H₉NOS
Storage Temperature: Ambient
MDL Number: MFCD18819336
CAS Number: 150079-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR