1-Benzylpiperazin-2-one 97%
Supplier: Apollo Scientific
OR912955-1G
OR912955-5G
APOSOR912955-1GEA
190
EUR
APOSOR912955-1G
APOSOR912955-5G
1-Benzylpiperazin-2-one 97%
1-Benzylpiperazin-2-one
Formula:
C₁₁H₁₄N₂O MW: 190.242 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD04038519 CAS Number: 59702-21-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...