1-Propylpyrrolidin-2-one 95%

Supplier: Apollo Scientific

OR913396-1G OR913396-25G OR913396-5G
APOSOR913396-1GEA 116 EUR
APOSOR913396-1G APOSOR913396-25G APOSOR913396-5G
1-Propylpyrrolidin-2-one 95%
1-Propylpyrrolidin-2-one
Formula: C₇H₁₃NO
MW: 127.184 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09749878
CAS Number: 3470-99-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR